missing translation for 'onlineSavingsMsg'
Learn More

Lorglumide sodium salt, 98+%, Thermo Scientific™

Product Code. p-7042210
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15494249 25 mg 25mg
15404259 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15494249 Supplier Thermo Scientific Alfa Aesar Supplier No. J61713.MB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and specific non-peptide cholecystokinin (CCK) receptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97964-56-2
Molecular Formula C22H32Cl2N2O4
Molecular Weight (g/mol) 459.41
MDL Number MFCD00083183
InChI Key IEKOTSCYBBDIJC-UHFFFAOYNA-N
Synonym lorglumide, lorglumide inn, lorglumidum latin, lorglumida spanish, ccris 6857, chembl24938, d,l-4-3,4-dichlorobenzoylamino-5-dipentylamino-5-oxo-pentanoic acid, lorglumide sodium salt, lorglumida, lorglumidum
PubChem CID 3960
IUPAC Name 4-[(3,4-dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxopentanoic acid
SMILES CCCCCN(CCCCC)C(=O)C(CCC(O)=O)NC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Color White
Quantity 25 mg
Formula Weight 481.39
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Lorglumide sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.