Learn More
Loratadine-d4 (piperidinylidene-3,3,5,5-d4), CDN
Specifications
Specifications
| Chemical Name or Material | Loratadine-D4 (piperidine-3,3,5,5-D4) |
| CAS | 2748435-73-4 |
| Molecular Formula | C22 D4 H19 Cl N2 O2 |
| Quantity | 0.005 g |
| Synonym | Ethyl 4-(8-chloro-5,6-dihydro-11 H-benzo[5,6 ]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinecarboxylate-3,3,5,5-d4, Loratadine-d4, Loratadine-D4 (piperidine-3,3,5,5-D4), Loratadine-d4 (piperidinylidene-3,3,5,5-d4) |
| InChI Formula | InChI=1 S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14 H,2,5-6,9-10,12-13H2,1H3/i9D2,10D2 |
| SMILES | [2 H]C1([2 H])CN(CC([2 H])([2 H])C1=C2c3ccc(Cl)cc3CCc4cccnc24)C(=O)OCC |
| IUPAC Name | ethyl 4-(13-chloro-4-azatricyclo[9.4.0.03,8 ]pentadeca-1(11),3(8),4,6,12,14-hexaen-2-ylidene)-3,3,5,5-tetradeuteriopiperidine-1-carboxylate |
| Molecular Weight (g/mol) | 386.9079 |
| Formula Weight | 386.1699 g/mol |
| Show More |