Learn More
Lonza™ GelBond™ PAG Film for Acrylamide Gels
Product Code. 11537765
Click to view available options
Quantity:
10/Pk.
50/Pk.
Length (English):
6.2 in.
7 in.
10.1 in.
10.2 in.
10.3 in.
Width (English):
4.8 in.
5.4 in.
6.3 in.
6.4 in.
7.8 in.
7.9 in.
13.7 in.
Unit Size:
10 sheets
50 sheets
Product Code. 11537765
Brand: Lonza™ 54729
Please to purchase this item. Need a web account? Register with us today!
Transparent, flexible polyester film supports polyacrylamide or MDE™ gels through electrophoresis, fixing, staining, destaining, and drying procedures
  • Covalently attaches to the acrylamide monomers during the polymerization reaction
  • Makes gels easier to handle
  • Helps gels retain their original dimensions during staining, destaining, and drying
TRUSTED_SUSTAINABILITY
Specifications
Gel Size 16.5 x 22 cm
Product Type PAG Film for Acrylamide Gels
Material Polyester
Quantity 50/Pk.
Thickness (English) 0.007 in.
Thickness (Metric) 0.02 cm
Length (English) 10.1 in.
Length (Metric) 22 cm
Width (English) 6.4 in.
Width (Metric) 22 cm