Learn More
Lonza™ AccuGENE™ 5M Sodium Chloride
Description
- Ready-to-use
- Prepared with 18megohm water
- Filtered using 0.2μ filter and filled into sterile bottles
- Manufactured according to strict quality control standards to ensure consistency
- Guaranteed DNase, RNase and protease-free
- Customized solutions are available to meet individual needs
Specifications
Specifications
| Concentration | 5 M |
| DNase | Yes |
| For Use With (Application) | Ideal for a wide range of molecular biology applications |
| Product Type | 5M Sodium Chloride |
| RNase | Yes |
| Quantity | 1000 mL |
For research or manufacturing use only.