Description
- Wipe away stubborn spills with our strong and absorbent cleaning cloths.
- Prevent cross-contamination by using different-coloured cloths for different jobs.
- No need to change your cleaning routine - these cloths can be used with most solvents, disinfectants and detergents.
- Avoid waste with strong and durable cloths that won't tear easily and can be reused.
Specifications
Specifications
| Product Type | Cleaning Cloth |
| Length (English) | 15.16 in. |
| Length (Metric) | 38.5 cm |
| Width (English) | 11.81 in. |
| Width (Metric) | 30 cm |
| Certifications/Compliance | FSC MIX Credit, ISO 9001, ISO 14001 |
| Color | Green |
| Packaging Quantity | 320/Cs. |
| Ply | 1-ply |
| Certifications/Compliance (Europe) | FSC MIX Credit, ISO 9001, ISO 14001 |