missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lomibuvir, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Lomibuvir (VX-222), a selective, non-nucleoside polymerase inhibitor, targets thumb pocket 2 of the HCV NS5B polymerase (RdRp) with a Kd of 17 nM. Lomibuvir inhibits the 1b/Con1 HCV subgenomic replicon with an EC50 of 5.2 nM. Lomibuvir preferentially inhibits elongative RNA synthesis rather than de novo-initiated RNA synthesis.
Specifications
Specifications
| Chemical Name or Material | Lomibuvir |
| CAS | 1026785-55-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H35NO4S |
| Quantity | 50 mg |
| Synonym | VX-222 |
| Solubility Information | DMSO : 100 mg/mL (224.41 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(N([C@@H]2CC[C@@H](O)CC2)C([C@H]3CC[C@H](C)CC3)=O)C=C(C#CC(C)(C)C)S1)O |
| Molecular Weight (g/mol) | 445.61 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction