Learn More
Lofexidine, MedChemExpress
Product Code. 16434537
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16434537 Supplier Medchem Express Supplier No. HYB1052A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Lofexidine is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal.
Lofexidine is a selective α2-receptor agonist, commonly used to alleviate the physical symptoms of heroin and other types of opioid withdrawal.
Specifications
Chemical Name or Material Lofexidine
CAS 31036-80-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12Cl2N2O
Quantity 50 mg
Solubility Information DMSO : 62.5 mg/mL (241.19 mM; Need ultrasonic)
SMILES CC(C1=NCCN1)OC2=C(Cl)C=CC=C2Cl
Molecular Weight (g/mol) 259.13
Formula Weight 259.13
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation