missing translation for 'onlineSavingsMsg'
Learn More

Lodoxamide, MedChemExpress

Product Code. 16436018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436018 10 mg 10mg
16486426 100 mg 100mg
16481116 200 mg 200mg
16478627 5 mg 5mg
16452157 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16436018 Supplier Medchem Express Supplier No. HY14270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lodoxamide (U-42585E free acid) is an antiallergic compound acting as a mast-cell stabilizer for the treatment of asthma and allergic conjunctivitis.

Lodoxamide (U-42585E free acid) is an antiallergic compound acting as a mast-cell stabilizer for the treatment of asthma and allergic conjunctivitis.

Specifications

Chemical Name or Material Lodoxamide
Physical Form Solid
CAS 53882-12-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H6ClN3O6
For Use With (Application) COVID-19-immunoregulation
Synonym U-42585E free acid
Solubility Information DMSO : 50 mg/mL (160.45 mM; Need ultrasonic)
SMILES OC(C(NC1=C(Cl)C(NC(C(O)=O)=O)=CC(C#N)=C1)=O)=O
Molecular Weight (g/mol) 311.63
Formula Weight 311.63
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.