Learn More
Lnx1 Antibody (4E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084708-M01
This item is not returnable.
View return policy
Description
Lnx1 Monoclonal antibody specifically detects Lnx1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| Lnx1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX | |
| LNX1 (NP_116011, 19 a.a. ∽ 118 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DNVGNLHFLYSELCKGAFHYGLTKDRKRRSQDGCPDGCASLTATAPSPEVSAAATISLMTDEPGLDNPAYVSSAEDGQPAISPVDSGRSNRTRARPFERS | |
| 0.1 mg | |
| Lipid and Metabolism, Zinc Finger | |
| 84708 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 4E3 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_116011 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |