missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMW-PTP/ACP1 Antibody (5C11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59404H
This item is not returnable.
View return policy
Description
LMW-PTP/ACP1 Monoclonal antibody specifically detects LMW-PTP/ACP1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| LMW-PTP/ACP1 | |
| Monoclonal | |
| HRP | |
| acid phosphatase 1, soluble, acid phosphatase of erythrocyte, Adipocyte acid phosphatase, cytoplasmic phosphotyrosyl protein phosphatase, EC 3.1.3.2, EC 3.1.3.48, HAAP, LMW-PTP, LMW-PTPase, Low molecular weight cytosolic acid phosphatase, low molecular weight phosphotyrosine protein phosphatase, MGC111030, MGC3499, protein tyrosine phosphatase, Red cell acid phosphatase 1 | |
| Recombinant human LMW-PTP/ACP1 (1-158aa) purified from E. coli | |
| 0.1 mL | |
| Protein Phosphatase | |
| 52 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 5C11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction