missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMO3 Polyclonal Antibody

Description
LMO3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene belongs to the rhombotin family of cysteine-rich LIM domain oncogenes. This gene is predominantly expressed in the brain. Related family members, LMO1 and LMO2 on chromosome 11, have been reported to be involved in chromosomal translocations in T-cell leukemia. Many alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Aug 2011].
Specifications
Specifications
| Antigen | LMO3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | LMO3 |
| Gene Accession No. | Q8TAP4 |
| Gene Alias | AI854781; BB106490; LIM domain only 3; LIM domain only 3 (rhombotin-like 2); LIM domain only protein 3; LIM only 3; Lmo3; LMO-3; Neuronal-specific transcription factor DAT1; RBTN3; Rbtn-3; RBTNL2; RGD1561357; RHOM3; Rhom-3; Rhombotin-3 |
| Gene Symbols | LMO3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?