Learn More
Lithocholic acid, 95%
Product Code. p-154033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571091 5 g Glass bottle 5g
10511354 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10571091 Supplier Thermo Scientific Acros Supplier No. 229090050
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Lithocholic acid, CAS # 434-13-9, a bacterial metabolite, is an oligo (ethylene glycol) and a secondary bile acid. | CAS: 434-13-9 | C24H40O3 | 376.581 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
  • Lithocholic acid is a monohydroxy acid, and a secondary bile acid
Applications
  • Lithocholic acid can be used in studies that assess cholestasis
  • It can be used to study hepatic phospholipid regulation and homeostasis of bile acid
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 434-13-9
Molecular Formula C24H40O3
Molecular Weight (g/mol) 376.581
MDL Number MFCD00003682
InChI Key SMEROWZSTRWXGI-HVATVPOCSA-N
Synonym lithocholic acid, lithocolic acid, lithocholate, 3alpha-hydroxy-5beta-cholanic acid, 3alpha-hydroxy-5beta-cholan-24-oic acid, 3-hydroxycholan-24-oic acid, 3alpha-hydroxycholanic acid, 3-alpha-hydroxycholanic acid, 5beta-cholanic acid-3alpha-ol, 3alpha-hydroxy-5beta-cholanoic acid
PubChem CID 9903
ChEBI CHEBI:16325
IUPAC Name (4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(CCC3C2CCC4C3(CCC(C4)O)C)C
Specifications
Melting Point 182°C to 188°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 10, IV, 785
Merck Index 15, 5601
Specific Rotation Condition + 34.00 (20.00°C c=1.5,C2H5OH)
Specific Rotation On dry basis: +32° to +36° (20°C, 589nm) (c=1.5, C2H5OH)
Formula Weight 376.57
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Lithocholic acid, 98%