missing translation for 'onlineSavingsMsg'
Learn More

Lithium tetraborate, Puratronic™, 99.998% (metals basis)

Product Code. p-4781976
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
This item is not returnable. View return policy

Product Code. 11326837

Brand: Thermo Scientific Alfa Aesar 010744.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol

Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2
Molecular Formula B4Li2O7
Molecular Weight (g/mol) 169.11
MDL Number MFCD00011083
InChI Key RDZCISUGRQUANA-UHFFFAOYSA-N
IUPAC Name dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Melting Point 930°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Moderately soluble in water.
Formula Weight 169.12
Density 2.4 g/cm3 at 20°C
Chemical Name or Material Lithium tetraborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.