missing translation for 'onlineSavingsMsg'
Learn More

Lithium hydride, 99.4% (metals basis)

Product Code. p-7051584
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11328728

Brand: Thermo Scientific Alfa Aesar 041596.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7580-67-8 | HLi | 7.948 g/mol

Lithium hydride is used as a precursor in the production of a variety of reagents in the synthesis, especially silane by reacting with silicon tetrachloride through Sundermeyer process and lithium aluminum hydride and lithium borohydride. It finds application in nuclear reactors as a shielding material. In addition, it is a hydrogen source or reducing agent to prepare other hydrides amides and 2H isotopic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7580-67-8
Molecular Formula HLi
Molecular Weight (g/mol) 7.948
MDL Number MFCD00011074
InChI Key SRTHRWZAMDZJOS-UHFFFAOYSA-N
Synonym lithium hydride, lithiumhydride, lih, lithium hydride anion, hydride lithium, hydridolithate 1-, lih-, ksc491a4n
PubChem CID 62714
IUPAC Name lithium;hydride
SMILES [H-].[Li+]

Specifications

Melting Point 686°C
Color White to Gray
Physical Form Powder
Quantity 25 g
Linear Formula LiH
UN Number UN1414
Merck Index 14,5533
Solubility Information Slightly soluble in dimethylformamide. Insoluble in acetone,benzene and toluene.
Formula Weight 7.95
Percent Purity 99.4%
Odor Odorless
Sensitivity Air and moisture sensitive
Density 0.78 g/mL
Assay (metals basis)
Chemical Name or Material Lithium hydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lithium hydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Lithium hydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.