missing translation for 'onlineSavingsMsg'
Learn More

Lithium benzoate, 99+%

Product Code. p-7023228
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11467897

Brand: Thermo Scientific Alfa Aesar A16571.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 553-54-8 | C7H5LiO2 | 128.06 g/mol

In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic electroluminescent devices, the device with lithium benzoate gives the best performance of all the devices studied. Aromatic multilayers of lithium benzoate were covalently attached to the surface of natural graphite through diazonium chemistry, and the resulting graphite exhibited superior electrochemical performance as anode material of lithium ion batteries.

Solubility
Solubility in water is increased by sodium benzoate

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-54-8
Molecular Formula C7H5LiO2
Molecular Weight (g/mol) 128.06
MDL Number MFCD00035540
InChI Key LDJNSLOKTFFLSL-UHFFFAOYSA-M
Synonym lithium benzoate, benzoic acid, lithium salt, benzoic acid lithium salt, unii-r9z042z19e, benzoic acid, lithium salt 1:1, lithium benzoate nf, lithiumbenzoate, lithotab benzoate, lithium 1+ ion benzoate, ksc491a5n
PubChem CID 2724073
SMILES [Li+].[O-]C(=O)C1=CC=CC=C1

Specifications

Melting Point >300°C
Odor Odorless
Quantity 250 g
Beilstein 3598089
Merck Index 14,5523
Solubility Information Solubility in water is increased by sodium benzoate
IUPAC Name lithium;benzoate
Formula Weight 128.06
Percent Purity ≥99%
Chemical Name or Material Lithium benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.