missing translation for 'onlineSavingsMsg'
Learn More

Lithium aluminum hydride, powder, 97%, Thermo Scientific™

Product Code. 11376409
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1000 g
Unit Size:
1000g
100g
10g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376409 100 g 100g
11356409 10 g 10g
11366409 25 g 25g
11486360 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11376409 Supplier Thermo Scientific Alfa Aesar Supplier No. A18116.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Lithium aluminum hydride is used as a reducing agent in various synthetic organic chemistry reactions. For example, it is used in the conversion of esters, carboxylic acids, acyl chlorides, aldehydes and ketones into their corresponding alcohols. It also involved in the conversion of amide, nitro, nitrile, imine, oxime and azide compounds into their amines as well as reduces alkyl halides to alkanes. It reacts with sodium chloride to prepare sodium hydride. It acts as a potential hydrogen storage medium, which could be used for future fuel cell-powered vehicles. It is mainly used in the preparation of other tetrahydridoaluminumates like sodium aluminum hydride, potassium aluminum hydride and magnesium alanate. Furthermore, it has been used to prepare thermoplastic polyester polyamides from oleic acid and lithium-polymer batteries.

Solubility
Soluble in water, diethyl ether and terahydrofuran.

Notes
Air and moisture sensitive. Incompatible with water, strong oxidizing agents, alcohols, carboxylic acid, peroxides, chlorinated solvents, halogens and oxygen. It reacts violently with water and release hydrogen gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16853-85-3
Molecular Formula AlH4Li
Molecular Weight (g/mol) 37.95
MDL Number MFCD00011075
InChI Key OCZDCIYGECBNKL-UHFFFAOYSA-N
Synonym lithium aluminum hydride, aluminum lithium hydride, lithiumaluminiumhydride, aluminum iii lithium hydride, lithium alanate, lithium aluminum tetrahydride, lithiumaluminiumhydrid, litiumaluminum hydride, lithim aluminum hydride
PubChem CID 21226445
IUPAC Name lithium(1+) alumanuide
SMILES [Li+].[AlH4-]

Specifications

Melting Point >125°C (decomposition)
Physical Form Powder
Quantity 100 g
UN Number UN1410
Merck Index 14,349
Solubility Information Soluble in water,diethyl ether and terahydrofuran.
Formula Weight 37.95
Percent Purity 97%
Odor Odorless
Sensitivity Moisture sensitive
Density 0.917
Chemical Name or Material Lithium aluminum hydride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.