missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIR-8/CD85c/LILRB5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98702-100ul
This item is not returnable.
View return policy
Description
LIR-8/CD85c/LILRB5 Polyclonal antibody specifically detects LIR-8/CD85c/LILRB5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LIR-8/CD85c/LILRB5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8CD85C, LIR-8leukocyte immunoglobulin-like receptor subfamily B member 5, member 5 | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human LIR-8/CD85c/LILRB5. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10990 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction