Learn More
Borer Chemie™ deconex™ 11 UNIVERSAL Liquid
For the manual and automatic cleaning of laboratory glassware, instruments and utensils. Borer Chemie™ deconex™ 11 UNIVERSAL Liquid provides residue-free removal of stubborn organic and inorganic soil.
Brand:  Borer Chemie™ 500100.00-F10W
Product Code. 10712721
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
  • Environmentally friendly
  • Mildly alkaline with phosphates
  • With active chlorine
  • Also for radioactive decontamination
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • deconex 11 UNIVERSAL
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.