missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIPI Polyclonal Antibody
GREENER_CHOICE

Product Code. 15957404
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15957404 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15957404 Supplier Invitrogen™ Supplier No. PA549416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIPI Polyclonal Antibody for Western Blot

Hydrolyzes specifically phosphatidic acid (PA) to produce 2-acyl lysophosphatidic acid (LPA; a potent bioactive lipid mediator) and fatty acid. Does not hydrolyze other phospholipids, like phosphatidylserine (PS), phosphatidylcholine (PC) and phosphatidylethanolamine (PE) or triacylglycerol (TG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPI
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene LIPI
Gene Accession No. Q6XZB0
Gene Alias cancer/testis antigen 17; CT17; D930038D03Rik; lipase I; lipase member I; lipase, member I; LIPI; LPD lipase; lpd1; LPDL; Membrane-associated phosphatidic acid-selective phospholipase A1-beta; mPA-PLA1 beta; PLA1C; PRED5
Gene Symbols LIPI
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 224-258 amino acids from the Central region of human LIPI.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149998
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.