Learn More
LIPG Monoclonal Antibody (OTI4C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501020S
Encompass_Preferred
 View more versions of this product
Product Code. 17533061
  • 242.00 € / 30µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
LIPG Monoclonal antibody specifically detects LIPG in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Specifications
Specifications
LIPG
Monoclonal
0.76 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y5X9
Mouse
Affinity chromatography
RUO
9388
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
OTI4C3
Unconjugated
LIPG
3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719
Full length human recombinant protein of human LIPG produced in HEK293T cell.
30 μL