missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipase A Antibody (9G7F12) [mFluor Violet 450 SE], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Lipase A Monoclonal antibody specifically detects Lipase A in Human samples. It is validated for ELISA,Western Blot,Serum
Specifications
Specifications
| Antigen | Lipase A |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 9G7F12 |
| Conjugate | mFluor Violet 450 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Acid cholesteryl ester hydrolase, Cholesteryl esterase, EC 3.1.1, EC 3.1.1.13, LALCESDcholesterol ester hydrolase, Lipase A, lipase A, lysosomal acid, cholesterol esterase, lysosomal acid lipase, lysosomal acid lipase/cholesteryl ester hydrolase, Sterol esterase |
| Host Species | Mouse |
| Immunogen | Truncated recombinant Lipase A expressed in E. Coli strain BL21 (DE3). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?