Learn More
Linerixibat, MedChemExpress
Product Code. 16493137
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16493137
Brand: Medchem Express HY16643
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Linerixibat (GSK2330672) is a highly potent, nonabsorbable and orally active apical sodium-dependent bile acid transporter (ASBT) inhibitor with an IC50 of 42 nM human ASBT. Linerixibat can be used as lipid-lowering agent. Linerixibat has the potential for type 2 diabetes and Primary Biliary Cholangitis treatment.
Linerixibat (GSK2330672) is a highly potent, nonabsorbable and orally active apical sodium-dependent bile acid transporter (ASBT) inhibitor with an IC50 of 42 nM human ASBT. Linerixibat can be used as lipid-lowering agent. Linerixibat has the potential for type 2 diabetes and Primary Biliary Cholangitis treatment.
Specifications
Chemical Name or Material Linerixibat
CAS 1345982-69-5
Color White
Purity Grade Notes Research
Molecular Formula C28H38N2O7S
Quantity 50 mg
Synonym GSK2330672
Solubility Information DMSO : 50 mg/mL (91.46 mM; Need ultrasonic)
SMILES O=C(O)CC(NCC1=C(OC)C=C(C2=C1)[C@@H](C3=CC=CC=C3)N[C@](CC)(CCCC)CS2(=O)=O)CC(O)=O
Molecular Weight (g/mol) 546.68
Formula Weight 546.68
Percent Purity 99.67%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism