Learn More
Invitrogen™ LIN28A Monoclonal Antibody (6D1F9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515453
Description
MA5-15453 targets LIN28 in indirect ELISA, IF, ICC, and WB applications and shows reactivity with human samples. The MA5-15453 immunogen is purified recombinant fragment of LIN28 (aa93-209) expressed in E. Coli. MA5-15453 detects LIN28 which has a predicted molecular weight of approximately 23kDa.
Lin28 is a transcription factor that was first identified through its key role in the pathway of developmental timing in C. elegans. The role of Lin28 in development suggested that it might be useful in the creation of stem cells that might be beneficial in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing Lin28 in addition to the transcription factors POU5F1, Sox2, and NANOG in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Lin28, Sox2, POU5F1 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
Specifications
| LIN28A | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide; pH 7.4 | |
| Q9H9Z2 | |
| LIN28A | |
| Purified recombinant fragment of LIN28 (aa93-209) expressed in E. Coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| 6D1F9 | |
| Unconjugated | |
| LIN28A | |
| AL024421; CSDD1; ENSMUSG00000070700; FLJ12457; Gm10299; Lin28; LIN-28; lin-28 homolog A; lin-28 homolog A (C. elegans); Lin28a; lin-28A; Protein lin-28 homolog A; RGD1566408; RNA-binding protein LIN-28; sb:cb650; si:ch211-232d9.4; similar to RNA-binding protein LIN-28; testis expressed gene 17; testis-expressed protein 17; Tex17; ZCCHC1; zgc:55584; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 79727 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.