missing translation for 'onlineSavingsMsg'
Learn More

LIN-41/TRIM71 Antibody, FITC, Novus Biologicals™

Product Code. 18555470 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18555470 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18555470 Supplier Novus Biologicals Supplier No. NBP176281F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIN-41/TRIM71 Polyclonal antibody specifically detects LIN-41/TRIM71 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN-41/TRIM71
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias LIN41, LIN-41, tripartite motif containing 71, tripartite motif-containing protein 71
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human TRIM71. The immunogen is located within amino acids 680 - 730 of TRIM71. Amino Acid Squence: CGEKGTKNGQFNYPWD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131405
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.