Learn More
LIMS2 Monoclonal Antibody (OTI5H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811408S
Description
LIMS2 Monoclonal antibody specifically detects LIMS2 in Human samples. It is validated for Western BlotSpecifications
| LIMS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5SQQ9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11023 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5H8 | |
| Unconjugated | |
| VAX1 | |
| ILK-binding protein; LGMD2W; LIM and senescent cell antigen like domains 2; LIM and senescent cell antigen-like domains 2; LIM and senescent cell antigen-like-containing domain protein 2; LIM zinc finger domain containing 2; LIM-like protein 2; Lims2; LIM-type zinc finger domains 2; Particularly interesting new Cys-His protein 2; Pinch2; PINCH-2 | |
| Full length human recombinant protein of human VAX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.