Learn More
LIMS2 Monoclonal Antibody (OTI5H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811408
Description
Adapter protein in a cytoplasmic complex linking beta- integrins to the actin cytoskeleton, bridges the complex to cell surface receptor tyrosine kinases and growth factor receptors. Plays a role in modulating cell spreading and migration.Specifications
| LIMS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z4I7, Q91XD2 | |
| Lims2 | |
| Human recombit protein fragment corresponding to amino acids 1-363 of human LIMS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5H8 | |
| Unconjugated | |
| LIMS2 | |
| LGMD2W, PINCH-2, PINCH2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 225341, 55679 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.