missing translation for 'onlineSavingsMsg'
Learn More

LIMS2 Antibody, Novus Biologicals™

Product Code. 18216882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18216882 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18216882 Supplier Novus Biologicals Supplier No. NBP152185

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

LIMS2 Polyclonal specifically detects LIMS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIMS2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001129509.2
Gene Alias FLJ10044, ILK-binding protein, LIM and senescent cell antigen-like domains 2, LIM and senescent cell antigen-like-containing domain protein 2, LIM-like protein 2, Particularly interesting new Cys-His protein 2, PINCH2, PINCH-2
Gene Symbols LIMS2
Host Species Goat
Immunogen Peptide with sequence C-RWSNMSDALA corresponding to internal region according to NP_001129509.2, NP_001154875.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55679
Test Specificity This antibody is expected to recognize isoform 1 (NP_001129509.2) and isoformn 3 (NP_001154875.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.