Learn More
Invitrogen™ Limitin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547798
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse IFN-gamma is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.01 EU per 1 μg of the antibody by the LAL method.
Has antiviral and antiproliferative activities. Produced by macrophages and stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| Limitin | |
| Polyclonal | |
| Unconjugated | |
| Ifna11 | |
| 6030405N23Rik; BGIF; IFN-[a]11; Ifna11; IFN-alpha-11; Ifnz; IFN-z; interferon alpha 11; interferon alpha family, gene 11; interferon alpha-11; interferon zeta; Limitin; Lmtn | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 15964 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q61716 | |
| Ifna11 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta Leu22-Arg182. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.