missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97870-100ul
This item is not returnable.
View return policy
Description
LIM Kinase 1 Polyclonal antibody specifically detects LIM Kinase 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LIM Kinase 1 | |
| Polyclonal | |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| Produced in rabbits immunized with E. coli-derived Human LIM Kinase 1 fragment. | |
| 100 μg | |
| Breast Cancer, Cancer, Protein Kinase | |
| 3984 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction