missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1 Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
LIM Kinase 1 Polyclonal antibody specifically detects LIM Kinase 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | LIM Kinase 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 510-647 of LIM Kinase 1 (NP_002305.1).,, Sequence:, VGNPYWMAPEMINGRSYDEKVDVFSFGIVLCEIIGRVNADPDYLPRTMDFGLNVRGFLDRYCPPNCPPSFFPITVRCCDLDPEKRPSFVKLEHWLETLRMHLAGHLPLGPQLEQLDRGFWETYRRGESGLPAHPEVPD |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?