Learn More
Epredia™ LifterSlip™ mSeries Cover Slips for Microarray Slides, approx. 20µL
Offer superior surface quality and flatness to reduce the variation in hybridization fluid volume across the microarray surface.
Specifications
Description mSeries LifterSlip Cover Slips for Microarray Slides
View More Specs
Description
Description
  • Precise ink-printed raised edges to provide consistent dispersal and easy injection of hybridization solution
  • Without adhesives to prevent leaching at high temperatures or extended hybridization times
Specifications
Specifications
mSeries LifterSlip Cover Slips for Microarray Slides
Videos
SDS
Documents
Documents
Product Certifications