Learn More
Epredia™ LifterSlip™ mSeries Cover Slips for Microarray Slides, approx. 20µL
Offer superior surface quality and flatness to reduce the variation in hybridization fluid volume across the microarray surface.
Description
- Precise ink-printed raised edges to provide consistent dispersal and easy injection of hybridization solution
- Without adhesives to prevent leaching at high temperatures or extended hybridization times
Specifications
| mSeries LifterSlip Cover Slips for Microarray Slides |