missing translation for 'onlineSavingsMsg'
Learn More

Lifitegrast, MedChemExpress

Product Code. p-200102153
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16497738

Brand: Medchem Express HY19344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lifitegrast (SAR 1118) is a potent integrin antagonist. Lifitegrast blocks the binding of intercellular adhesion molecule 1 (ICAM-1) to lymphocyte function-associated antigen 1 (LFA-1), interrupting the T cell-mediated inflammatory cycle. Lifitegrast inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Lifitegrast can be used for researching dry eye disease.

Lifitegrast (SAR 1118) is a potent integrin antagonist. Lifitegrast blocks the binding of intercellular adhesion molecule 1 (ICAM-1) to lymphocyte function-associated antigen 1 (LFA-1), interrupting the T cell-mediated inflammatory cycle. Lifitegrast inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Lifitegrast can be used for researching dry eye disease.

Specifications

Chemical Name or Material Lifitegrast
CAS 1025967-78-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H24Cl2N2O7S
Quantity 10 mg
Synonym SAR 1118 SHP-606
Solubility Information DMSO : ≥ 29 mg/mL (47.12 mM)
SMILES O=C(O)[C@H](CC1=CC=CC(S(=O)(C)=O)=C1)NC(C2=C(Cl)C3=C(CN(C(C4=CC=C5C=COC5=C4)=O)CC3)C=C2Cl)=O
Molecular Weight (g/mol) 615.48
Formula Weight 615.48
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.