Learn More
Lificiguat, MedChemExpress
Product Code. 16476596
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16476596
Brand: Medchem Express HY14927
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Lificiguat binds to the β subunit of soluble guanylyl cyclase(sGC) with Kd of 0.6-1.1 μM in the presence of CO.
Lificiguat binds to the β subunit of soluble guanylyl cyclase(sGC) with Kd of 0.6-1.1 μM in the presence of CO.
Specifications
Chemical Name or Material Lificiguat
Physical Form Solid
CAS 170632-47-0
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H16N2O2
For Use With (Application) Cancer-Kinase/protease
Synonym YC-1
Solubility Information DMSO : ≥ 100 mg/mL (328.58 mM)
SMILES OCC1=CC=C(C2=NN(CC3=CC=CC=C3)C4=C2C=CC=C4)O1
Molecular Weight (g/mol) 304.34
Formula Weight 304.34
Percent Purity 98.82%
Grade Research