missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Imager UV Light Base

Product Code. 12393583
Click to view available options
For Use With (Equipment):
E-Gel Imager
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12393583

Brand: Invitrogen™ 4466602

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

1 x E-Gel™ Imager UV Light Base

E-Gel Imager UV Light Base has a built-in safety mechanism that requires the E-Gel Imager Camera Hood to be affixed over the top in order to be operational

The E-Gel Imager UV Light Base is one of three interchangeable bases that work with the E-Gel Imager Camera Hood to comprise the E-Gel Imager system. The E-Gel Imager UV Light base is a compact UV light transilluminator for self-poured gels as well as E-Gel™ agarose gel cassettes stained with ethidium bromide. UV Light Base is best for ethidium bromide.

  • E-Gel Adaptor Base has been designed to provide power to either E-Gel iBase power system coupled to E-Gel Safe Imager™, or E-Gel Go Base with its integrated blue light transilluminator so E-Gel cassettes can be run right inside E-Gel Imager
  • This allows for real time image capture while samples are running on the gel

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x E-Gel™ Imager UV Light Base.

Store at room temperature.
For Use With (Equipment) E-Gel Imager
Product Line E-Gel
Type Imager UV Light Base
Label or Dye Ethidium bromide
Shipping Condition Room Temperature
System Type E-Gel™ Imager

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.