missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LHR Antibody (LHCGR/7399) [Alexa Fluor™ 488], Novus Biologicals™

Product Code. 30066417 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30066417 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066417 Supplier Novus Biologicals™ Supplier No. NBP326966AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LHR Monoclonal antibody specifically detects LHR in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen LHR
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LHCGR/7399
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor
Host Species Mouse
Immunogen Recombinant fragment (around aa 100-300) of human LHR protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Caveolae Markers, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3973
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.