Learn More
LHF-535, MedChemExpress
Product Code. 16497407
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16497407
Brand: Medchem Express HY112762
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively.
LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively.
Specifications
Chemical Name or Material LHF-535
Physical Form Solid
CAS 1450929-77-7
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H28N2O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 96.67 mg/mL (234.34 mM; Need ultrasonic)
SMILES OC(C)(C)C1=CC=C(/C=C\C2=CC=C3C(N=CN3C4=CC=C(OC(C)C)C=C4)=C2)C=C1
Molecular Weight (g/mol) 412.52
Formula Weight 412.52
Percent Purity 99.2%
Grade Research