missing translation for 'onlineSavingsMsg'
Learn More

LHF-535, MedChemExpress

Product Code. 16460987
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460987

Brand: Medchem Express HY112762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively.

LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively.

Specifications

Chemical Name or Material LHF-535
Physical Form Solid
CAS 1450929-77-7
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C27H28N2O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 96.67 mg/mL (234.34 mM; Need ultrasonic)
SMILES OC(C)(C)C1=CC=C(/C=C\C2=CC=C3C(N=CN3C4=CC=C(OC(C)C)C=C4)=C2)C=C1
Molecular Weight (g/mol) 412.52
Formula Weight 412.52
Percent Purity 99.2%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.