missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ LHC Basal Medium (1X)
GREENER_CHOICE

Product Code. p-7076028
Click to view available options
Quantity:
1,000 mL
500 mL
Unit Size:
1000mL
500mL
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 11524556

Brand: Gibco™ 12677019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

LHC Basal Medium (1X)

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience, we offer these products with or without supplementation.

LHC Basal Medium (1X), liquid

Applications:

Formulated for the growth of bronchial epithelial cells with further supplements.

Recommended storage conditions:

2°C to 8°C.

Shelf life:

12 months

Intended use(s)

Research use only

LHC-8 Medium and LHC-9 Medium

Applications:

Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.

Performance and quality testing:

Performance tested in a growth assay using BEAS-2B cells.

Recommended storage conditions:

-5°C to -20°C.

Intended use(s)

Research use only

Reference(s):

Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line B3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells)
Cell Type Epithelial Cells (Bronchial)
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type LHC Basal Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Quantity 500 mL
Shipping Condition Room Temperature
Species Cattle/Bovine, Human
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.