Learn More
LGR5 Monoclonal Antibody (OTI14E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503155S
Description
LGR5 Monoclonal antibody specifically detects LGR5 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| LGR5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15785 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10953 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI14E1 | |
| Unconjugated | |
| Tomm34 | |
| Fex; follicle expressed hormone; G protein-coupled receptor 49; GPR49; GPR67; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; GRP49; HG38; leucine rich repeat containing G protein coupled receptor 5; leucine rich repeat containing G protein-coupled receptor 5; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; leucine-rich repeat-containing G-protein coupled receptor 5; Lgr5; orphan G protein-coupled receptor HG38; orphan G-protein coupled receptor FEX | |
| Full length human recombinant protein of human TOMM34 produced in HEK293 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.