Learn More
Tocris Bioscience™ LFM-A13
GREENER_CHOICE
Product Code. 18005262
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18005262
Brand: Tocris Bioscience™ 1300/10
Please to purchase this item. Need a web account? Register with us today!
Potent, selective BTK inhibitor
Potent and selective inhibitor of Bruton's tyrosine kinase (BTK). Inhibits recombinant BTK with an IC50 value of 2.5 μM and has no activity on other protein kinases ( including JAK1, JAK3, HCK, EGFR kinase and insulin receptor kinase) at concentrations of up to 278 μM.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62004-35-7
Molecular Formula C11H8Br2N2O2
Specifications
Quantity 10 mg
Formula Weight 360
Purity >98%
Molecular Weight (g/mol) 360.005
Chemical Name or Material LFM-A13