missing translation for 'onlineSavingsMsg'
Learn More

LFM-A13, MedChemExpress

Product Code. 16447448
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16447448

Brand: Medchem Express HY18009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibits recombinant BTK, Plx1 and PLK3 with IC50s of 2.5 μM, 10 μM and 61 μM; LFM-A13 shows no effects on JAK1 and JAK3, Src family kinase HCK, EGFR and IRK.

LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibits recombinant BTK, Plx1 and PLK3 with IC50s of 2.5 μM, 10 μM and 61 μM

Specifications

Chemical Name or Material LFM-A13
CAS 244240-24-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8Br2N2O2
Quantity 10 mg
Solubility Information DMSO : ≥ 42 mg/mL (116.67 mM)
SMILES BrC1=C(NC(/C(C#N)=C(C)\O)=O)C=C(Br)C=C1
Molecular Weight (g/mol) 360.0
Formula Weight 360.0
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.