Learn More
LFM-A13, MedChemExpress
Product Code. 16493467
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16493467
Brand: Medchem Express HY18009
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibits recombinant BTK, Plx1 and PLK3 with IC50s of 2.5 μM, 10 μM and 61 μM; LFM-A13 shows no effects on JAK1 and JAK3, Src family kinase HCK, EGFR and IRK.
LFM-A13 is a potent BTK, JAK2, PLK inhibitor, inhibits recombinant BTK, Plx1 and PLK3 with IC50s of 2.5 μM, 10 μM and 61 μM
Specifications
Chemical Name or Material LFM-A13
CAS 244240-24-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H8Br2N2O2
Quantity 50 mg
Solubility Information DMSO : ≥ 42 mg/mL (116.67 mM)
SMILES BrC1=C(NC(/C(C#N)=C(C)\O)=O)C=C(Br)C=C1
Molecular Weight (g/mol) 360.0
Formula Weight 360.0
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease