Learn More
Levosimendan Cyanoacetamide Hydrazone Impurity, TRC
Specifications
Specifications
| Chemical Name or Material | Levosimendan Cyanoacetamide Hydrazone Impurity |
| CAS | 274263-65-9 |
| Molecular Formula | C14H14N6O2 |
| Quantity | 25 mg |
| Synonym | 2-Cyano-2-[[4-[(4R)-1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazono]-acetamide, 2-Cyano-2-[2-[4-[(4R)-1,4,5,6-tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazinylidene]-acetamide |
| InChI Formula | InChI=1S/C14H14N6O2/c1-8-6-12(21)19-20-13(8)9-2-4-10(5-3-9)17-18-11(7-15)14(16)22/h2-5,8,17H,6H2,1H3,(H2,16,22)(H,19,21)/b18-11+/t8-/m1/s1 |
| SMILES | O=C1C[C@@H](C)C(C2=CC=C(N/N=C(C(N)=O)\C#N)C=C2)=NN1 |
| IUPAC Name | (1E)-2-amino-N-[4-[(4R)-4-methyl-6-oxo-4,5-dihydro-1H-pyridazin-3-yl]anilino]-2-oxoethanimidoyl cyanide |
| Molecular Weight (g/mol) | 298.3 |
| Formula Weight | 298.1178 |