Learn More
Levofloxacin Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Levofloxacin Hydrochloride |
| CAS | 177325-13-2 |
| Molecular Formula | C18H20FN3O4 . (HCl) |
| Quantity | 1 g |
| Synonym | Weilisu, (S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid Monohydrochloride, (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-Pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid Monohydrochloride |
| InChI Formula | InChI=1S/C18H20FN3O4.ClH/c1-10-9-26-17-14-11(16(23)12(18(24)25)8-22(10)14)7-13(19)15(17)21-5-3-20(2)4-6-21;/h7-8,10H,3-6,9H2,1-2H3,(H,24,25);1H/t10-;/m0./s1 |
| SMILES | Cl.C[C@H]1COc2c(N3CCN(C)CC3)c(F)cc4C(=O)C(=CN1c24)C(=O)O |
| Molecular Weight (g/mol) | 397.8 |
| Formula Weight | 397.1205 |