missing translation for 'onlineSavingsMsg'
Learn More

Levobupivacaine hydrochloride, MedChemExpress

Product Code. p-200100914
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451436 200 mg 200mg
16448406 100 mg 100mg
16434437 50 mg 50mg
16418568 10 mg 10mg
16402196 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16451436 Supplier Medchem Express Supplier No. HYB0653A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Levobupivacaine hydrochloride ((S)-(-)-Bupivacaine monohydrochloride) is a long-acting amide local anaesthetic.

Levobupivacaine hydrochloride exerts anaesthetic and analgesic effects through reversible blockade of neuronal sodium channel. Levobupivacaine hydrochloride can inhibit impulse transmission and conduction in cardiovascular and other tissues, possessing certain cardiac and CNS toxicity. Levobupivacaine hydrochloride is metabolized by hepatic cytochrome P450 (CYP450) enzymes in vivo. Levobupivacaine hydrochloride can also induce ferroptosis by miR-489-3p/SLC7A11 signaling in gastric cancer.

Specifications

Chemical Name or Material Levobupivacaine hydrochloride
CAS 27262-48-2
Color White
Purity Grade Notes Research
Molecular Formula C18H29ClN2O
Quantity 200 mg
Synonym (S)-(-)-Bupivacaine monohydrochloride
Solubility Information DMSO : 100 mg/mL (307.80 mM; Need ultrasonic)
SMILES O=C([C@H]1N(CCCC)CCCC1)NC2=C(C)C=CC=C2C.[H]Cl
Molecular Weight (g/mol) 324.89
Formula Weight 324.89
Percent Purity 99.37%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.