missing translation for 'onlineSavingsMsg'
Learn More

Levobunolol Hydrochloride, TRC

Product Code. 30134479
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134479 1 g 1g
30134173 250 mg 250mg
30011118 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134479 Supplier Toronto Research Chemicals Supplier No. TRCL3350001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27912-14-7
Molecular Formula C17 H25 N O3 . Cl H
Molecular Weight (g/mol) 327.85
Synonym Levobunolol Hydrochloride, 1(2H)-Naphthalenone, 5-[(2S)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (1:1), 1(2H)-Naphthalenone, 5-[(2S)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (9CI), 1(2H)-Naphthalenone, 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (-)- (8CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (S)-, AG 901, Betagan, Gotensin, Levobunolol hydrochloride, Vitagan, W 7000A, l-Bunolol hydrochloride
IUPAC Name 5-[(2S)-3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-2H-naphthalen-1-one;hydrochloride
SMILES Cl.CC(C)(C)NC[C@H](O)COc1cccc2C(=O)CCCc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H25NO3.ClH/c1-17(2,3)18-10-12(19)11-21-16-9-5-6-13-14(16)7-4-8-15(13)20;/h5-6,9,12,18-19H,4,7-8,10-11H2,1-3H3;1H/t12-;/m0./s1
Formula Weight 327.1601
Percent Purity >95%
Chemical Name or Material Levobunolol, Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.