Learn More
Levobunolol Hydrochloride, TRC
Product Code. 30134173
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134173 250 mg 250mg
30134479 1 g 1g
30011118 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134173 Supplier Toronto Research Chemicals Supplier No. TRCL335000250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27912-14-7
Molecular Formula C17 H25 N O3 . Cl H
Molecular Weight (g/mol) 327.85
Synonym Levobunolol Hydrochloride, 1(2H)-Naphthalenone, 5-[(2S)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (1:1), 1(2H)-Naphthalenone, 5-[(2S)-3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride (9CI), 1(2H)-Naphthalenone, 5-[3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (-)- (8CI), 1(2H)-Naphthalenone, 5-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]-3,4-dihydro-, hydrochloride, (S)-, AG 901, Betagan, Gotensin, Levobunolol hydrochloride, Vitagan, W 7000A, l-Bunolol hydrochloride
IUPAC Name 5-[(2S)-3-(tert-butylamino)-2-hydroxypropoxy]-3,4-dihydro-2H-naphthalen-1-one;hydrochloride
SMILES Cl.CC(C)(C)NC[C@H](O)COc1cccc2C(=O)CCCc12
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C17H25NO3.ClH/c1-17(2,3)18-10-12(19)11-21-16-9-5-6-13-14(16)7-4-8-15(13)20;/h5-6,9,12,18-19H,4,7-8,10-11H2,1-3H3;1H/t12-;/m0./s1
Formula Weight 327.1601
Percent Purity >95%
Chemical Name or Material Levobunolol, Hydrochloride
Grade HPLC