Learn More
Levobetaxolol hydrochloride, MedChemExpress
Product Code. 16464347
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464347 50 mg 50mg
16438468 10 mg 10mg
16438276 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16464347 Supplier Medchem Express Supplier No. HYB0381B
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor (beta blocker) that can lower the pressure in the eye.
Levobetaxolol hydrochloride is a beta-adrenergic receptor inhibitor (beta blocker) that can lower the pressure in the eye.
Specifications
Chemical Name or Material Levobetaxolol hydrochloride
CAS 116209-55-3
Color White
Purity Grade Notes Research
Molecular Formula C18H30ClNO3
Quantity 50 mg
Synonym (S)-Betaxolol hydrochloride AL-1577A
Solubility Information DMSO : ≥ 100 mg/mL (290.79 mM) ∣H2O : 25 mg/mL (72.70 mM; Need ultrasonic)
SMILES O[C@@H](CNC(C)C)COC1=CC=C(CCOCC2CC2)C=C1.[H]Cl
Molecular Weight (g/mol) 343.89
Formula Weight 343.89
Percent Purity 98.53%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation