missing translation for 'onlineSavingsMsg'
Learn More

Levetiracetam, MedChemExpress

Product Code. 16468096
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16468096

Brand: Medchem Express HYB0106

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.

Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.

Specifications

Chemical Name or Material Levetiracetam
CAS 102767-28-2
Color White
Purity Grade Notes Research
Quantity 100 mg
Synonym UCB L059
Solubility Information H2O : ≥ 85 mg/mL (499.38 mM) ∣DMSO : ≥ 85 mg/mL (499.38 mM)
SMILES NC([C@@H](N(CCC1)C1=O)CC)=O
Molecular Weight (g/mol) 170.21
Percent Purity 99.21%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.