Learn More
Levetiracetam, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Description
Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.
Specifications
Specifications
| Chemical Name or Material | Levetiracetam |
| CAS | 102767-28-2 |
| Color | White |
| Purity Grade Notes | Research |
| Quantity | 50 mg |
| Synonym | UCB L059 |
| Solubility Information | H2O : ≥ 85 mg/mL (499.38 mM) ∣DMSO : ≥ 85 mg/mL (499.38 mM) |
| SMILES | NC([C@@H](N(CCC1)C1=O)CC)=O |
| Molecular Weight (g/mol) | 170.21 |
| Percent Purity | 99.21% |
| Show More |